Learn More
anti-Histone H3, Clone: mAbcam 24834, Abcam™
This product is for research use only and not for diagnostic or therapeutic procedures. Customer is granted a limited, non-exclusive license for internal research and shall not attempt to reverse engineer.
Supplier: Abcam AB24834100UG

Description
Host Species: Mouse
Clone: mAbcam 24834
Isotype: IgG3, κ
Cross Reactivity: Human. Predicted to work with a wide range of other species, all Mammals.
Immunogen: Synthetic peptide derived from within residues 100 to the C-terminus of Human Histone H3. Generated with the same immunogen as our successful rabbit polyclonal ab1791
Applications:
Western Blotting
Specifications
Histone H3 | |
mAbcam 24834 | |
Unconjugated | |
Synthetic peptide derived from within residues 100 to the C-terminus of Human Histone H3. Generated with the same immunogen as our successful rabbit polyclonal ab1791 | |
Epigenetics And Nuclear Signaling | |
Human, Mammalia |
Monoclonal | |
1mg/mL | |
Mouse | |
100 μg | |
Primary | |
IgG3 κ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.