Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3 Mouse anti-Human, Drosophila, Clone: 6HH3-2C5, MilliporeSigma™

Catalog No. 051354MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-135-4MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-135-4MI Supplier MilliporeSigma Supplier No. 051354
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Use Anti-Histone H3 Antibody (Dimethyl K9, Phospho S10), clone 6HH3-2C5 (mouse monoclonal antibody) validated in western blot, immunocytochemistry to detect Histone H3 (Dimethyl K9 also known as H3K9me2S10P, Histone H3 (di methyl K9 phospho S10).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H3 (Dimethyl K9, Phospho S10)
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 6HH3-2C5
Conjugate Unconjugated
Formulation Mouse monoclonal IgG1κ ascites with 0.05% sodium azide.
Gene Accession No. AK304607
Gene Symbols H3.3B, H3F3A, H3F3B
Host Species Mouse
Immunogen Synthetic linear peptide corresponding to dimethyl (Lys9) and Phospho (Ser10) of Histone H3.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Target Species Drosophila, Human
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.