Learn More
Histone H4 (citrulline 3) Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Histone H4 (citrulline 3) |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Gene Symbols | HIST1H4A; H4/J; HIST1H4F; HIST1H4L; H4FN; H4FA; H4FH; HIST1H4H; H4F2; H4FM; H4/A; H4FK; H4FG; HIST2H4; H4FB; H4/K; HIST1H4I; H4/N; HIST1H4B; H4FE; H4/M; H4/a; HIST1H4E; HIST4H4; HIST2H4A; H4FC; H4FI; H4/H; HIST1H4C; H4/G; HIST1H4K; H4FJ; H4FD; H4/I; H4/B; H4/C; H4/D; HIST1H4J; HIST1H4D; H4/E |
| Host Species | Rabbit |
| Immunogen | synthetic peptide corresponding to amino acids 1-10 of human histone H4, where [cit] corresponds to the change of arginine to citrulline at residue 3 |
| Purification Method | Affinity Purified |
| Quantity | 200 μL |
| Primary or Secondary | Primary |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.