Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone-lysine N-methyltransferase SUV39H2 Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABE530
Description
Detect SUV39H1 using this rabbit polyclonal antibody, Anti-Histone-lysine N-methyltransferase SUV39H2 Antibody validated for use in western blotting and immunoprecipitation.
Specifications
Histone-lysine N-methyltransferase SUV39H2 | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human |
Immunoprecipitation, Western Blot | |
Unconjugated | |
SUV39H2; KMT1B | |
KLH-conjugated linear peptide corresponding to human Histone-lys N-methyltransferase SUV39H2. | |
100 μg | |
NP_001180355.1 | |
2-8°C for 12 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction