Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone Macro H2A.1 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. ABE215MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE215MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE215MI Supplier MilliporeSigma Supplier No. ABE215
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Histone Macro H2A.1 Antibody is a Rabbit Polyclonal Antibody for detection of Histone Macro H2A.1 also known as H2A histone family member Y, histone macroH2A1.2 and has been validated in western blot, immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone Macro H2A.1
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols H2A.y; H2AF12M; H2AFJ; H2A/y; MACROH2A1.1; mH2A; MACROH2A
Host Species Rabbit
Immunogen GST-tagged recombinant protein corresponding to mcro domain of mouse Histone MacroH2A.1.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_001152985.1
Target Species Human, Mouse
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.