Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIV Mouse anti-Human, Clone: 7A8.1, MilliporeSigma™

Catalog No. MAB880AMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB880AMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB880AMI Supplier MilliporeSigma Supplier No. MAB880A
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-HIV Antibody, p24, clone 7A8.1 is an antibody against HIV for use in IC and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIV
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 7A8.1
Conjugate Unconjugated
Formulation Purified. Supplied in 0.05M Tris Cl, pH 8.0.
Host Species Mouse
Immunogen p24 isolated from human (native) HIV infected cells.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C for up to 12 months
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.