Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-HIV-I p17, Clone: 32/5.8.42, ZeptoMetrix

Murine monoclonal antibody reacts with Human Immunodeficiency Virus Type 1 (HIV-1) p17 protein
$410.00 - $2583.00
Specifications
| Antigen | HIV-I p17 |
|---|---|
| Clone | 32/5.8.42 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
22-157-316
![]() |
ZeptoMetrix
0801005 |
100 μg |
Each for $410.00
|
|
|||||
22-157-353
![]() |
ZeptoMetrix
0801123 |
1 mg |
Each for $2,583.00
|
|
|||||
Description
Host Species: Mouse
Clone: 32/5.8.42
Isotype: IgG2a
Species Reactivity: Hu
Enzyme Immunoassay, Immunofluorescence, Western Blotting
Specifications
| HIV-I p17 | |
| Monoclonal | |
| Purified | |
| RUO | |
| IgG2a |
| 32/5.8.42 | |
| Unconjugated | |
| Mouse | |
| Human | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title