Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-HIV-I p24, Clone: 32/5.17.76, ZeptoMetrix

Murine monoclonal antibody reacts with Human Immuno-deficiency Virus Type 1 (HIV-1) p24 protein
$347.00 - $2440.00
Specifications
Antigen | HIV-I p24 |
---|---|
Clone | 32/5.17.76 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
22-157-330
![]() |
ZeptoMetrix
0801078 |
100 μg |
Each for $347.00
|
|
|||||
22-157-354
![]() |
ZeptoMetrix
0801124 |
1 mg |
Each for $2,440.00
|
|
|||||
Description
Host Species: Mouse
Clone: 32/5.17.76
Isotype: IgG1
Species Reactivity: Hu
Enzyme Immunoassay, Immunofluorescence, Western Blotting
Specifications
HIV-I p24 | |
Monoclonal | |
Purified | |
RUO | |
IgG1 |
32/5.17.76 | |
Unconjugated | |
Mouse | |
Human | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title