Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-HIV-I p24, Clone: 39/6.14, ZeptoMetrix
SDP

Catalog No. 22157391 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-157-391 1 mg
22-157-333 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-157-391 Supplier ZeptoMetrix Supplier No. 0802014
Add to Cart
Add to Cart

Murine monoclonal antibody reacts with Human Immunodeficiency Virus Type 1 (HIV - 1) p24 protein

Host Species: Mouse
Clone: 39/6.14
Isotype: IgG1
Species Reactivity: Hu

Enzyme Immunoassay, Immunofluorescence, Western Blotting

Specifications

Antigen HIV-I p24
Classification Monoclonal
Clone 39/6.14
Conjugate Unconjugated
Host Species Mouse
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.