Learn More
Description
Host Species: Mouse
Clone: 38/8.7.47
Isotype: IgG1
Species Reactivity: Hu
Enzyme Immunoassay, Immunofluorescence, Western Blotting
Specifications
Specifications
| Antigen | HIV-I p24 |
| Classification | Monoclonal |
| Clone | 38/8.7.47 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Quantity | 1 mg |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Target Species | Human |
| Form | Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
