Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HIV1 gp41 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA17219
Description
PA1-7219 detects the HIV gp41. This antibody does not cross-react with human T or B cells. PA1-7219 has been successfully used in Western blot procedures. The PA1-7219 immunogen is recombinant (P. Pastoris) ectodomain of gp41 (glycosylated).
Gp41 is a glycoprotein, that along with gp120 is integral to the process by which HIV enters the cell. Gp41 is primarily located within the viral envelope, however when gp120 binds to a CD4 receptor, gp120 changes its conformation, causing gp41 to become.
Specifications
| HIV1 gp41 | |
| Polyclonal | |
| Unconjugated | |
| HIV Glycoprotein 41 | |
| Recombinant ectodomain of gp41-glycosylated (a.a. residues 546-682) from strain HxB2. | |
| 1 mL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 5 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Goat | |
| IgG Fraction | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction