Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIV1 nef Monoclonal Antibody (3F2), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA171500
Description
This antibody reacts with HIV-1 subtype C Nef of BRU, IIIB, LAV and HAN strains. It has been mapped to amino acids 31-35 (-GAASR-, HIV-1 HXB2).
HIV1 nef Monoclonal antibody specifically detects HIV1 nef in Virus samples. It is validated for ELISA, Western Blot.Specifications
HIV1 nef | |
Monoclonal | |
0.8 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
Mouse | |
Protein G | |
RUO | |
Virus | |
Antibody | |
IgG1 |
ELISA, Western Blot | |
3F2 | |
Unconjugated | |
HIV-1 Nef | |
Recombinant Nef protein of HIV-1 HXB2 purified from E. coli | |
100 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction