Learn More
HK2, Mouse, Clone: 4H1, Abnova™
Mouse monoclonal antibody raised against a partial recombinant HK2.
Supplier: Abnova Corporation H00003099M01
Description
Hexokinases phosphorylate glucose to produce glucose-6-phosphate, the first step in most glucose metabolism pathways. This gene encodes hexokinase 2, the predominant form found in skeletal muscle. It localizes to the outer membrane of mitochondria. Expression of this gene is insulin-responsive, and studies in rat suggest that it is involved in the increased rate of glycolysis seen in rapidly growing cancer cells. [provided by RefSeq
Sequence: IVKEVCTVVARRAAQLCGAGMAAVVDRIRENRGLDALKVTVGVDGTLYKLHPHFAKVMHETVKDLAPKCDVSFLQSEDGSGKGAALITAVACRIREAGQRSpecifications
HK2 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC021116 | |
HK2 | |
HK2 (AAH21116, 818 a.a. ∼ 917 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG2a κ |
ELISA, Western Blot | |
4H1 | |
Mouse monoclonal antibody raised against a partial recombinant HK2. | |
HK2 | |
DKFZp686M1669/HKII/HXK2 | |
Mouse | |
Affinity chromatography | |
RUO | |
3099 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.