Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-A2 Mouse anti-Human, PE, Clone: BB7.2, BD

Catalog No. BDB558570 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558570 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558570 Supplier BD Biosciences Supplier No. 558570
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The monoclonal antibody BB7.2 reacts with a subunit of the human leukocyte antigen-A2 (HLA-A2), a class I molecule of the major histocompatibility complex (MHC). The MHC gene locus encodes a group of highly polymorphic, cell-surface proteins that play a broad role in the immune response to protein antigens. MHC molecules function by binding and presenting small antigenic protein fragments to antigen-specific receptors expressed by T cells (TCR). Human (human leukocyte antigen/HLA) MHC molecules comprise two major classes, MHC class I and class II. Functionally, class I MHC molecules can bind peptides derived from intracellular antigens (eg, viral and some bacterial antigens) that are specifically recognized by CD8+ T cells, and class II MHC molecules recognize antigens derived from pathogens multiplying in intracellular vesicles, and those derived from ingested extracellular bacteria. When presented on the cell surface by the MHC class II molecules, these antigens are recognized by CD4+ T cells. TCR recognize both processed peptides bound to MHC, as well as regions of the MHC molecule itself. CD4 and CD8 accessory molecules strengthen formation of the TCR-MHC complex through their interaction with non-polymorphic regions of the MHC molecule.

Blocking Assay, Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA-A2
Applications Flow Cytometry
Classification Monoclonal
Clone BB7.2
Conjugate PE
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.