Learn More
HLA-ABC Mouse anti-Human, Alexa Fluor 488, Clone: DX17, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 560169
Description
The DX17 monoclonal antibody reacts with a monomorphic epitope expressed on all HLA (human leukocyte antigen) class I molecules examined. DX17 immunoprecipitates HLA class I heavy chains (45kDa) and β2-microglobulin (12kDa) from radiolabeled human cell lines. HLA is determined by a complex segment of the short arm of chromosome 6 and there are many human HLAs encoded in this segment. The antigenic agglomerate is called MHC, for major histocompatibility complex. Examples of class I loci are HLA-A, -B, and -C, which are serologically assayed; class II loci, e.g., HLA-D/DR and DC1, are tested by lymphocytotoxic methods.
Host Species: Mouse
Clone: DX18
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Polyclonal Human NK Cell Line
Flow Cytometry

Specifications
HLA-ABC | |
Monoclonal | |
Alexa Fluor 488 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
DX17 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Polyclonal Human NK Cell Line | |
100 Tests | |
Immunology | |
Human | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.