Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-ABC Common Mouse anti-Human, Clone: 22.63.4, MilliporeSigma™

Catalog No. MAB1275MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1275MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1275MI Supplier MilliporeSigma Supplier No. MAB1275
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-HLA-ABC Common Antibody, clone 22.63.4 is validated for use in flow cytometry, immunocytochemistry, immunohistochemistry for the detection of HLA-ABC.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA-ABC Common
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry
Classification Monoclonal
Clone 22.63.4
Conjugate Unconjugated
Formulation Purified from mouse ascitic fluid. The antibody is supplied in phosphate buffered saline, pH 7.4 containing 0.1% sodium azide, diluted 50% v/v with glycerol. The characteristics of each lot are tested by electrophoresis and flow cytometry.
Gene Accession No. NM_002116.5
Gene Symbols HLA-A, A-1, Aw-34, A-10, Aw-74, Aw-19, A-28, Aw-80, Aw-69, Aw-36, HLAA, Aw-66, Aw-43, A-9, Aw-68, Aw-24, Aw-33
Host Species Mouse
Immunogen Nalm-6 cell line
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.