Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-ABC Common Mouse anti-Human, FITC, Clone: 22.63.4, MilliporeSigma™

Catalog No. MAB1275FMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1275FMI 1 Unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1275FMI Supplier MilliporeSigma Supplier No. MAB1275F

Mouse Monoclonal Antibody

Detect HLA-ABC using this Anti-HLA-ABC Common Antibody, clone 22.63.4, FITC conjugated validated for use in FC and immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA-ABC Common
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone 22.63.4
Conjugate FITC
Formulation Purified from mouse ascitic fluid. Fluorescein conjugated. The antibody is supplied in 1.0ml phosphate buffered saline, pH 7.4, containing 0.2% bovine serum albumin and 0.1% sodium azide. The characteristics of each lot are tested by electrophoresis and
Gene Accession No. NM_002116.5
Gene Symbols HLA-A, A-1, Aw-34, A-10, Aw-74, Aw-19, A-28, Aw-80, Aw-69, Aw-36, HLAA, Aw-66, Aw-43, A-9, Aw-68, Aw-24, Aw-33
Host Species Mouse
Immunogen Nalm-6 cell line
Quantity 1 Unit
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for 6 months. Do not freeze. Protect from light.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.