Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-A,B,C Mouse anti-Human, FITC, Clone: G46-2.6, BD

Catalog No. BDB555552 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
50 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555552 100 Tests
BDB557348 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB555552 Supplier BD Biosciences Supplier No. 555552
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with a monomorphic epitope of major histocompatibility Class I antigens widely distributed on human nucleated cells. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: G46-2.6
Isotype: IgG1 κ
Species Reactivity: Human, Baboon/Rhesus/Cynomolgus

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA-A,B,C
Applications Flow Cytometry
Classification Monoclonal
Clone G46-2.6
Conjugate FITC
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Baboon, Cynomolgus, Human, Rhesus Monkey
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.