Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-ABC Mouse anti-Human, PE, Clone: DX17, BD

Catalog No. BDB560168 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560168 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560168 Supplier BD Biosciences Supplier No. 560168
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The DX17 monoclonal antibody reacts with a monomorphic epitope expressed on all HLA (human leukocyte antigen) class I molecules examined. DX17 immunoprecipitates HLA class I heavy chains (45kDa) and β2-microglobulin (12kDa) from radiolabeled human cell lines. HLA is determined by a complex segment of the short arm of chromosome 6 and there are many human HLAs encoded in this segment. The antigenic agglomerate is called MHC, for major histocompatibility complex. Examples of class I loci are HLA-A, -B, and -C, which are serologically assayed; class II loci, e.g., HLA-D/DR and DC1, are tested by lymphocytotoxic methods.

Host Species: Mouse
Clone: DX17
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Polyclonal Human NK Cell Line

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA-ABC
Applications Flow Cytometry
Classification Monoclonal
Clone DX17
Conjugate PE
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Polyclonal Human NK Cell Line
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.