Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-B7 Mouse anti-Human, Phycoerythrin, Clone: BB7.1, MilliporeSigma™

Catalog No. MAB1288MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1288MI 1 Unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1288MI Supplier MilliporeSigma Supplier No. MAB1288
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-HLA-B7 Antibody, clone BB7.1, PE Conjugate is validated for use in FC for the detection of HLA-B7.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA-B7
Applications Flow Cytometry
Classification Monoclonal
Clone BB7.1
Conjugate PE
Formulation Purified mouse immunoglobulin in phosphate buffered saline, pH 7.4, containing 0.2% bovine Serum albumin and 0.1% sodium azide.
Gene Accession No. NM_005514.5
Gene Symbols HLA-B, Bw-54, Bw-45, Bw-47, Bw-46, Bw-52, B-5, Bw-4, Bw-57, OTTHUMP00000034826, AS, Bw-56, Bw-55, Bw-41, Bw-48, B', DT, Bw-50, Bw-60, Bw-58, B-21, HLA-B-5501, B-12, Bw-22, Bw-53, HLAB, SPDA1, Bw-44
Host Species Mouse
Immunogen Papain solubilized HLA-B7 antigen1
Purification Method Protein A purfied
Quantity 1 Unit
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.