Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-DM Mouse anti-Human, Unlabeled, Clone: MAP.DM1, BD

Catalog No. BDB555982 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555982 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555982 Supplier BD Biosciences Supplier No. 555982
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with human leukocyte antigen-DM (HLA-DM), a non-classical MHC class II molecule expressed in the cytoplasm of antigen presenting cells (APC). HLA-DM is composed of alpha and beta subunits which form a similar structure as the classical class II molecules. HLA-DM catalyzes the dissociation of CLIP from MHC class II-CLIP complexes in vitro and facilitates the binding of antigenic peptides. It also prevents self-antigens from becoming stably complexed with class II molecules and being presented to T cells.

Host Species: Mouse
Clone: MaP.DM1
Isotype: IgG1 κ
Species Reactivity: Human

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA-DM
Applications Flow Cytometry
Classification Monoclonal
Clone MAP.DM1
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.