Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-DQ Mouse anti-Human, FITC, Clone: SK10, BD

Catalog No. BDB347453 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB347453 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB347453 Supplier BD Biosciences Supplier No. 347453
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The Anti–HLA-DQ antibody, clone SK10, is derived from the hybridization of NS-1 mouse myeloma cells with spleen cells isolated from BALB/c mice immunized with tonsillar B lymphocytes (non–E-rosetting cells).

Anti-HLA-DQ (Leu-10) recognizes a class II major histocompatibility complex (MHC) antigen, Mr 26 to 34 kdaltons (kDa), distinct from HLA-DR and HLA-DP antigens. Anti-HLA-DQ recognizes a common polymorphic epitope present on HLA-DQ molecules of cells expressing DQw1 and DQw3 (usually associated with DR1, DR2, DR4, DR5, DRw8, DRw9, and DRw10) and absent from DQw2 (usually associated with DR3, and DR7). Therefore, Anti-HLA-DQ will not react with cells from DR3 or DR 7 homozygotes or DR3/7 heterozygotes.

TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA-DQ
Applications Flow Cytometry
Classification Monoclonal
Clone SK10
Concentration 25μg/mL
Conjugate FITC
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.