Learn More
HLA-DQ Mouse anti-Human, PE-CF594, Clone: Tu169, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 564807
Description
The TU169 monoclonal antibody specifically binds to the human Major Histocompatibility Complex (MHC) Class II HLA-DQ1 and DQ2 antigens and weakly to HLA-DQ3. HLA-DQ antigens exits as heterodimers comprised of polymorphic transmembrane HLA-DQ alpha and beta glycoproteins. HLA-DQ is expressed primarily on B cells, monocytes, macrophages, dendritic cells and activated T lymphocytes. HLA-DQ is involved in presenting peptidic antigens to CD4+ T lymphocytes. This antibody does not cross-react with HLA-DR2 and HLA-DR7. This antibody fixes complement.
This antibody is conjugated to BD Horizon PE-CF594, which has been developed exclusively by BD Biosciences as a better alternative to PE-Texas Red™. PE-CF594 excites and emits at similar wavelengths to PE-Texas Red™ yet exhibits improved brightness and spectral characteristics. Due to PE having maximal absorption peaks at 496nm and 564nm, PE-CF594 can be excited by the blue (488-nm), green (532-nm) and yellow-green (561-nm) lasers and can be detected with the same filter set as PE-Texas Red™ (eg, 610/20-nm filter).
Specifications
| HLA-DQ | |
| Monoclonal | |
| PE-CF594 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG2a κ |
| Flow Cytometry | |
| Tu169 | |
| MHC class II HLA-DQ; MHC HLA-DQ | |
| Mouse | |
| 100 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.