Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-DR Mouse anti-Human, Alexa Fluor 700, Clone: L243 (G46-6), BD

Catalog No. BDB560743 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560743 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560743 Supplier BD Biosciences Supplier No. 560743
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with HLA-DR, a human class II antigen II of the major histocompatibility complex (MHC). HLA-DR is a transmembrane glycoprotein composed of an α chain (36kD) and a β subunit (27kD) expressed primarily on antigen presenting cells: B cells, monocytes, macrophages, and thymic epithelial cells. HLA-DR is also expressed on activated T cells. This molecule plays a major role in cellular interaction during antigen presentation.

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA-DR
Applications Flow Cytometry
Classification Monoclonal
Clone L243 (G46-6)
Conjugate Alexa Fluor 700
Description MHC class II antigen; HLA class II histocompatibility antigen
Formulation Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.