Learn More
HLA-DR Mouse anti-Human, APC, Clone: L243 (G46-6), BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 340549
Description
Antin++DR recognizes a human class II major histocompatibility complex (MHC) antigen. The antigen is a transmembrane glycoprotein composed of α- and β-subunits that have molecular weights of 36 and 27kDa, respectively. Antin++DR reacts with a nonpolymorphic HLA-DR epitope and does not cross-react with HLA-DQ or HLA-DP molecules.
Flow Cytometry

Specifications
HLA-DR | |
Monoclonal | |
50μg/mL | |
Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
100 Tests | |
Immunology | |
Human | |
IgG2a κ |
Flow Cytometry | |
L243 (G46-6) | |
APC | |
Mouse | |
RUO | |
Primary | |
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.