Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-DR Mouse anti-Human, FITC, Clone: 169-1B5.1, MilliporeSigma™

Catalog No. MAB4201FMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4201FMI 1 Unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB4201FMI Supplier MilliporeSigma Supplier No. MAB4201F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect HLA-DR using this Anti-HLA-DR Antibody, clone 169-1B5.1, FITC conjugated validated for use in flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA-DR
Applications Flow Cytometry
Classification Monoclonal
Clone 169-1B5.1
Conjugate FITC
Formulation The antibody is supplied in 1.0ml phosphate buffered saline, pH 7.4, containing 0.2% bovine serum albumin and 0.1% sodium azide. The characteristics of each lot are tested by electrophoresis and flow cytometry.
Gene Accession No. NM_001025158.1; NM_001025159.1; NM_004355.2
Gene Symbols HLA-DR, p33, DHLAG, Ia-GAMMA, HLADG, HLA-DR-gamma
Host Species Mouse
Immunogen Human peripheral blood leukocytes.
Quantity 1 Unit
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for 6 months. Do not freeze. Protect from light.
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.