Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

high mobility group AT-hook 2, Mouse, Clone: 2D10, Abnova™

Catalog No. 89014218 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-014-218 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-014-218 Supplier Abnova Corporation Supplier No. H00008091M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant HMGA2.

This gene encodes a protein that belongs to the nonhistone chromosomal high mobility group (HMG) protein family. HMG proteins function as architectural factors and are essential components of the enhancesome. This protein contains structural DNA-binding domains and may act as a transcriptional regulating factor. Identification of the deletion, amplification, and rearrangement of this gene that are associated with myxoid liposarcoma suggests a role in adipogenesis and mesenchymal differentiation. A gene knock out study of the mouse counterpart demonstrated that this gene is involved in diet-induced obesity. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq

Sequence: MSARGEGAGQPSTSAQGQPAAPAPQKRGRGRPRKQQQEPTGEPSPKRPRGRPKGSKNKSPSKAAQKKAEATGEKRPRGRPRKWPQQVVQKKP

Specifications

Antigen high mobility group AT-hook 2
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 2D10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant HMGA2.
Formulation In 1X PBS, pH 7.4
Gene HMGA2
Gene Accession No. NM_003483
Gene Alias BABL/HMGI-C/HMGIC/LIPO/STQTL9
Gene Symbols HMGA2
Host Species Mouse
Immunogen HMGA2 (NP_003474, 1 a.a. ∼ 92 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8091
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.