Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HMGB1 Hamster anti-Mouse, Human, Rat, Clone: HMG1-5H6, MilliporeSigma™

Catalog No. MABE148MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE148MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE148MI Supplier MilliporeSigma Supplier No. MABE148
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

Use Anti-HMGB1 Antibody, clone HMG1-5H6 (Hamster Monoclonal Antibody) validated in western blot, immunofluoresence, FC to detect HMGB1 also known as High-mobility group box 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HMGB1
Applications Flow Cytometry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone HMG1-5H6
Conjugate Unconjugated
Formulation Purified hamster monoclonal in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_034569
Gene Symbols HMGB1, HMG1, Amphoterin, HMG3, SBP-1
Host Species Hamster
Immunogen Recombinant protein corresponding to mouse HMGB1.
Purification Method Protein G
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.