Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-HNE-Michael Adducts, Reduced, Polyclonal

Catalog No. 393207100UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
39-320-7100UL 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 39-320-7100UL Supplier MilliporeSigma™ Supplier No. 393207100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects HNE-Michael Adducts, Reduced Clone: in Many Species samples, and it is validated for ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immuno Blotting

The CNS is enriched in polyunsaturated fatty acids and is therefore particularly vulnerable to lipid peroxidation. 4-Hydroxy-2-noneal (HNE) is one of the major lipid peroxidation products with cytotoxic and mutagenic activity. It reacts with protein residues such as histidine to generate stable Michael adducts. HNE has been shown to inhibit neurite outgrowth and disrupt microtubules and tubulin modification, suggesting it may play a role in Alzheimer's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HNE-Michael Adducts, Reduced
Applications ELISA, Immunoblot, Immunoblot, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation Serum diluted in 50% glycerol, pH 7.5.
Host Species Rabbit
Immunogen HNE-treated KLH in which 1:1 amino acid-HNE Michael adducts were chemically reduced
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes chemically reduced amino acid-(4)-HNE adducts. Does not recognize non-reduced adducts or reduced modifications arising from malondialdehyde (MDA) and structurally-related alkenes such astrans-2-noneal.
Target Species Multi-species
Content And Storage ≤ -70°C
Form Diluted
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.