Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HNF-1α Mouse, Unlabeled, Clone: 2, BD

Catalog No. BDB610903 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610903 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610903 Supplier BD Biosciences Supplier No. 610903

Mouse Monoclonal Antibody

HNF-1 (Hepatocyte Nuclear Factor 1), a POU-homeodomain containing transcription factor, is a family member of liver-enriched transcription factors that also includes C-EBP, HNF-3, and HNF-4. Although HNF-1 exhibits elevated expression in liver and is thought to be involved in liver organogenesis, it is also found in kidney, stomach, intestine, and at a low level in thymus and spleen. A short N-terminal α-helical segment allows for HNF-1 homodimerization which is followed by DNA binding via an atypical helix-turn-helix motif. The HNF-1 dimer transactivates hepatic genes such as α- and β-fibrinogen, α-fetoprotein, albumin, α-L-antitrypsin, and pyruvate kinase by binding to a
highly conserved 13 bp inverted palindromic sequence, GTTAATNATTAAC. HNF-1 represses its own transcription and data suggests that there is coordinate regulation between HNF-1 and HNF-4. Although HNF-4 induces HNF-1 transcription, HNF-1 downregulates HNF-4 activity by binding to the AF2 region in the activation domain of HNF-4. The ability of HNF-1 to dictate the production or activity of itself and other transcription factors indicates its possible function as a general cellular control mechanism.

TRUSTED_SUSTAINABILITY

Specifications

Antigen HNF-1α
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone 2
Concentration 250μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Mouse HNF-1 aa. 70-269
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rabbit, Rat
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.