Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hepatocyte nuclear factor 4, alpha, Mouse, Clone: 3C6, Abnova™

Catalog No. 89000722 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-722 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-722 Supplier Abnova Corporation Supplier No. H00003172M07
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant HNF4A.

The protein encoded by this gene is a nuclear transcription factor which binds DNA as a homodimer. The encoded protein controls the expression of several genes, including hepatocyte nuclear factor 1 alpha, a transcription factor which regulates the expression of several hepatic genes. This gene may play a role in development of the liver, kidney, and intestines. Mutations in this gene have been associated with monogenic autosomal dominant noninsulin-dependent diabetes mellitus type I. Alternative splicing of this gene results in multiple transcript variants. [provided by RefSeq

Sequence: NDRQYDSRGRFGELLLLLPTLQSITWQMIEQIQFIKLFGMAKIDNLLQEMLLGGSPSDAPHAHHPLHPHLMQEHMGTNVIVANTMPTHLSNGQMCEWPRP

Specifications

Antigen hepatocyte nuclear factor 4, alpha
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 3C6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant HNF4A.
Formulation PBS with no preservative; pH 7.4
Gene HNF4A
Gene Accession No. NM_000457
Gene Alias FLJ39654/HNF4/HNF4a7/HNF4a8/HNF4a9/MODY/MODY1/NR2A1/NR2A21/TCF/TCF14
Gene Symbols HNF4A
Host Species Mouse
Immunogen HNF4A (NP_000448, 324 a.a. ∼ 423 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Stem Cell Biology
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 3172
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.