Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hnRNP A1 Mouse anti-Human, Clone: 9H10, MilliporeSigma™

Catalog No. 041469MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-146-9MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-146-9MI Supplier MilliporeSigma Supplier No. 041469
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-hnRNP A1 Antibody, clone 9H10 is a Mouse Monoclonal Antibody for detection of hnRNP A1 also known as Helix-destabilizing protein or Single-strand RNA-binding protein and has been validated in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen hnRNP A1
Applications Western Blot
Classification Monoclonal
Clone 9H10
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2bκ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Accession No. NP_002127
Gene Symbols HNRNPA1, HNRPA1, hnRNPA1
Host Species Mouse
Immunogen Recombinant protein corresponding to human hnRNP A1.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.