Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hnRNP L Mouse anti-Human, Clone: 4D11, MilliporeSigma™

Catalog No. 051518MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-151-8MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-151-8MI Supplier MilliporeSigma Supplier No. 051518

Mouse Monoclonal Antibody

Use Anti-hnRNP L Antibody, clone 4D11 (Mouse Monoclonal Antibody) validated in western blot, immunoprecipitation to detect hnRNP L also known as heterogeneous nuclear ribonucleoprotein L.
TRUSTED_SUSTAINABILITY

Specifications

Antigen hnRNP L
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 4D11
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_001005335
Gene Symbols HNRNPL, HNRPL, hnRNP L
Host Species Mouse
Immunogen Human hnRNP proteins (from HeLa cells) purified by affinity chromatography on ssDNA agarose.
Purification Method Protein G
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.