Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hnRNP-P2 Mouse anti-Mouse, Clone: LHC7.15, MilliporeSigma™

Catalog No. MABE192MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE192MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE192MI Supplier MilliporeSigma Supplier No. MABE192

Mouse Monoclonal Antibody

Anti-hnRNP-P2 Antibody, clone LHC7.15 is a Mouse Monoclonal Antibody for detection of hnRNP-P2 also known as RNA-binding protein FUS, Protein pigpen, hnRNP-P2 and has been validated in western blot, immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen hnRNP-P2
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone LHC7.15
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_631888
Gene Symbols Fus
Host Species Mouse
Immunogen Mouse spleen fused with SP2 cells
Purification Method Protein G purified
Quantity 200 μL
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.