Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HO-1/HMOX1/HSP32 Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.50 - $499.50

Specifications

Antigen HO-1/HMOX1/HSP32
Concentration 1.16 mg/mL
Dilution Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:25, Immunohistochemistry-Paraffin 1:200
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP177459SS
SDP
View Documents
Novus Biologicals
NBP177459SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NBP177459
SDP
View Documents
Novus Biologicals
NBP177459
0.1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

HO-1/HMOX1/HSP32 Polyclonal specifically detects HO-1/HMOX1/HSP32 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

HO-1/HMOX1/HSP32
Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:25, Immunohistochemistry-Paraffin 1:200
Polyclonal
Rabbit
Apoptosis, Cellular Markers, ER Markers, Lipid and Metabolism
PBS and 30% Glycerol with 0.05% Sodium Azide
bK286B10, EC 1.14.99.3, heat shock protein, 32-kD, heme oxygenase (decycling) 1, HO, HO1, HO-1heme oxygenase 1, HSP32
HMOX1
IgG
Affinity Purified
1.16 mg/mL
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human, Mouse
P09601
3162
A synthetic peptide made to an internal portion of the human Heme Oxygenase 1 protein (between residues 100-150) [UniProt# P09601]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.