Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOP Mouse anti-Human, Clone: 17E8.1, MilliporeSigma™

Catalog No. MABS182MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS182MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS182MI Supplier MilliporeSigma Supplier No. MABS182

Mouse Monoclonal Antibody

This HOP antibody is validated for use in western blotting for the detection of the HOP protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOP
Applications Western Blot
Classification Monoclonal
Clone 17E8.1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG3κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_006810
Gene Symbols STIP1
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human HOP.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG3 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.