Learn More
homeobox A1, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human HOXA1 protein.
Supplier: Abnova Corporation H00003198B01P
Description
In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. This gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation. The encoded protein may be involved in the placement of hindbrain segments in the proper location along the anterior-posterior axis during development. Two transcript variants encoding two different isoforms have been found for this gene, with only one of the isoforms containing the homeodomain region. [provided by RefSeq
Sequence: MDNARMNSFLEYPILSSGDSGTCSARAYPSDHRITTFQSCAVSANSCGGDDRFLVGRGVQIGSPHHHHHHHHHHPQPATYQTSGNLGVSYSHSSCGPSYGSQNFSAPYSPYALNQEADVSGGYPQCAPAVYSGNLSSPMVQHHHHHQGYAGGAVGSPQYIHHSYGQEHQSLALATYNNSLSPLHASHQEACRSPASETSSPAQTFDWMKVKRNPPKTGKVGEYGYLGQPNAVRTNFTTKQLTELEKEFHFNKYLTRARRVEIAASLQLNETQVKIWFQNRRMKQKKREKEGLLPISPATPPGNDEKAEESSEKSSSSPCVPSPGSSTSDTLTTSHSpecifications
homeobox A1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human HOXA1 protein. | |
HOXA1 | |
BSAS/HOX1/HOX1F/MGC45232 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Immunofluorescence, Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_005522.4 | |
HOXA1 | |
HOXA1 (NP_005513.1, 1 a.a. ∼ 335 a.a) full-length human protein. | |
50 μg | |
Transcription Regulation | |
3198 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.