Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOXA13 Mouse anti-Human, Rat, Mouse, Clone: 11E12.1, MilliporeSigma™

Catalog No. MABE155MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE155MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE155MI Supplier MilliporeSigma Supplier No. MABE155

Mouse Monoclonal Antibody

Use Anti-HOXA13 Antibody, clone 11E12.1 (Mouse Monoclonal Antibody) validated in western blotting to detect HOXA13 also known as Homeobox protein Hox-A13.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXA13
Applications Western Blot
Classification Monoclonal
Clone 11E12.1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.
Gene Accession No. NP_000513
Gene Symbols HOXA13, HOX1J
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human HOXA13.
Purification Method Ion Exchange
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.