Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HoxA7 Rabbit anti-Human, Vertebrates, Polyclonal, MilliporeSigma™

Catalog No. 09086MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-086-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-086-MI Supplier MilliporeSigma Supplier No. 09086

Rabbit Polyclonal Antibody

Anti-HoxA7 Antibody is a Rabbit Polyclonal Antibody for detection of HoxA7 also known as homeobox A7, Hox1.1 and has been validated in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HoxA7
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Immunoaffinity purified rabbit IgG in 0.1M Tris-Glycine (pH7.4), 15mM NaCl, 0.05% sodium azide.
Host Species Rabbit
Immunogen KLH-conjugated synthetic peptide<br/>corresponding to a.a. 29-41 of human HoxA7, synthetic peptide
corresponding to a.a. 29-41 of human HoxA7
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3204
Target Species Human, Vertebrates
Content And Storage 2-8°C for 12 months For maximum recovery of product centrifuge the vial prior to removing the cap.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.