Learn More
HOXB1, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human HOXB1 peptide using ARM Technology.
Supplier: Abnova Corporation H00003211K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, located on different chromosomes, consisting of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXB genes located in a cluster on chromosome 17. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
HOXB1 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
HOXB1 | |
HOXB1 | |
A synthetic peptide of human HOXB1 | |
RUO | |
3211 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
homeobox B1 | |
HOXB1 | |
HOX2, HOX2I, Hox-2.9, MGC116843, MGC116844, MGC116845 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.