Learn More
HOXB5, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human HOXB5 peptide using ARM Technology.
Supplier: Abnova Corporation H00003215K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene is a member of the Antp homeobox family and encodes a nuclear protein with a homeobox DNA-binding domain. It is included in a cluster of homeobox B genes located on chromosome 17. The encoded protein functions as a sequence-specific transcription factor that is involved in lung and gut development. Increased expression of this gene is associated with a distinct biologic subset of acute myeloid leukemia (AML) and the occurrence of bronchopulmonary sequestration (BPS) and congenital cystic adenomatoid malformation (CCAM) tissue. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
HOXB5 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
HOXB5 | |
HOXB5 | |
A synthetic peptide of human HOXB5 | |
RUO | |
3215 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
homeobox B5 | |
HOXB5 | |
HHO.C10, HOX2, HOX2A, HU-1, Hox2.1 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.