Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

homeobox C4, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017732 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-732 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-732 Supplier Abnova Corporation Supplier No. H00003221D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human HOXC4 protein.

This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, which are located on different chromosomes and consist of 9 to 11 genes arranged in tandem. This gene, HOXC4, is one of several homeobox HOXC genes located in a cluster on chromosome 12. Three genes, HOXC5, HOXC4 and HOXC6, share a 5' noncoding exon. Transcripts may include the shared exon spliced to the gene-specific exons, or they may include only the gene-specific exons. Two alternatively spliced variants that encode the same protein have been described for HOXC4. Transcript variant one includes the shared exon, and transcript variant two includes only gene-specific exons. [provided by RefSeq]

Sequence: MIMSSYLMDSNYIDPKFPPCEEYSQNSYIPEHSPEYYGRTRESGFQHHHQELYPPPPPRPSYPERQYSCTSLQGPGNSRGHGPAQAGHHHPEKSQSLCEPAPLSGASASPSPAPPACSQPAPDHPSSAASKQPIVYPWMKKIHVSTVNPNYNGGEPKRSRTAYTRQQVLELEKEFHYNRYLTRRRRIEIAHSLCLSERQIKIWFQNRRMKWKKDHRLPNTKVRSAPPAGAAPSTLSAATPGTSEDHSQSATPPEQQRAEDITRL

Specifications

Antigen homeobox C4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human HOXC4 protein.
Formulation PBS with no preservative; pH 7.4
Gene HOXC4
Gene Accession No. NM_014620.4
Gene Alias HOX3/HOX3E/cp19
Gene Symbols HOXC4
Host Species Rabbit
Immunogen HOXC4 (NP_055435.2, 1 a.a. ∼ 264 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Transcription Regulation
Primary or Secondary Primary
Gene ID (Entrez) 3221
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.