Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOXC8 Rabbit anti-Human, Polyclonal, Invitrogen™

Catalog No. PIPA527979
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA527979 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA527979 Supplier Thermo Scientific Supplier No. PA527979

Rabbit Polyclonal Antibody

PA5-27979 targets HOXC8 in WB applications and shows reactivity with Human samples. The PA5-27979 immunogen is recombit fragment corresponding to a region within amino acids 1 and 225 of Human HOXC8. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, which are located on different chromosomes and consist of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXC genes located in a cluster on chromosome 12. The product of this gene may play a role in the regulation of cartilage differentiation. It could also be involved in chondrodysplasias or other cartilage disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXC8
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Gene HOXC8
Gene Accession No. P31273
Gene Alias HOX3, HOX3A, Hox-3.1, mouse, homolog of, homeo box 3A, homeo box C8, homeobox protein Hox-3A, homeobox protein Hox-C8
Gene Symbols HOXC8
Host Species Rabbit
Immunogen Recombit fragment corresponding to a region within amino acids 1 and 225 of Human HOXC8
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3224
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.