Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

homeobox D1 (A01), Mouse anti-Human, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89004154 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-154 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-154 Supplier Abnova Corporation Supplier No. H00003231A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant HOXD1.

This gene is a member of the Antp homeobox family and encodes a protein with a homeobox DNA-binding domain. This nuclear protein functions as a sequence-specific transcription factor that is involved in differentiation and limb development. Mutations in this gene have been associated with severe developmental defects on the anterior-posterior (a-p) limb axis. [provided by RefSeq

Sequence: QVDYAAFGEPGPFPACLKASADGHPGAFQTASPAPGTYPKSVSPASGLPAAFSTFEWMKVKRNASKKGKLAEYGAASPSSAIRTNFSTKQ

Specifications

Antigen homeobox D1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant HOXD1.
Formulation 50% glycerol
Gene HOXD1
Gene Accession No. NM_024501
Gene Alias HOX4/HOX4G/Hox-4.7
Gene Symbols HOXD1
Host Species Mouse
Immunogen HOXD1 (NP_078777, 151 a.a. ∼ 240 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Transcription Regulation
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 3231
Target Species Human, Rat
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.