Learn More
HOXD3, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human HOXD3 peptide using ARM Technology.
Supplier: Abnova Corporation H00003232K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, located on different chromosomes, consisting of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXD genes located at 2q31-2q37 chromosome regions. Deletions that removed the entire HOXD gene cluster or 5' end of this cluster have been associated with severe limb and genital abnormalities. The protein encoded by this gene may play a role in the regulation of cell adhesion processes. [provided by RefSeq]
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
HOXD3 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
HOXD3 | |
HOXD3 | |
A synthetic peptide of human HOXD3 | |
RUO | |
3232 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
homeobox D3 | |
HOXD3 | |
HOX1D, HOX4, HOX4A, Hox-4.1, MGC10470 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.