Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HP1α Mouse anti-Human, Mouse, Vertebrates, Clone: 15.19s2, MilliporeSigma™

Catalog No. 05689MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-689-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-689-MI Supplier MilliporeSigma Supplier No. 05689
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-HP1α Antibody, clone15.19s2 is a Mouse Monoclonal Antibody for detection of HP1α also known as HP1Hs alpha, Heterochromatin protein 1 homolog alpha and has been validated in Chimmunoprecipitation, immunocytochemistry, immunohistochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HP1α
Applications ChIP Assay, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 15.19s2
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG in buffer containing 0.02Mphosphate buffer, pH 7.6, 0.25MNaCl, and 0.1% Sodium Azide before the addition of glycerol to 30%.
Gene Accession No. NM_012117
Gene Symbols CBX5, HP1A, HP1Hs-alpha, HP1-ALPHA, HP1
Host Species Mouse
Immunogen GST fusion protein corresponding to residues 2-191 of human HP1α protein.
Purification Method Protein G chromatography
Quantity 200 μg
Primary or Secondary Primary
Target Species Human, Mouse, Vertebrates
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.