Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HP1γ Mouse anti-Human, Mouse, Clone: 42s2, MilliporeSigma™

Catalog No. 05690MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-690-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-690-MI Supplier MilliporeSigma Supplier No. 05690
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Use Anti-HP1γ Antibody, clone 42s2 (Mouse Monoclonal Antibody) validated in Chimmunoprecipitation, immunohistochemistry, western blotting to detect HP1γ also known as Heterochromatin protein 1 homolog gamma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HP1γ
Applications ChIP Assay, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 42s2
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1 in buffer containing 0.014Mphosphate buffer, pH 7.6, 0.175M NaCl, 0.07% sodium azide, and 30% glycerol. Liquid at −20deg;C.
Gene Accession No. NP_009207
Gene Symbols CBX3, HECH, HP1-GAMMA, HP1Hs-gamma
Host Species Mouse
Immunogen GST fusion protein corresponding to residues 17-173 of human HP1γ protein. Clone 42s2.
Purification Method Protein G purfied
Quantity 200 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage −20°C for up to 12 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.