Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HPa2, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA521326
Description
PA5-21326 targets HPA2 in IHC (P) and WB applications and shows reactivity with Human and mouse samples.The PA5-21326 immunogen is a recombinant fragment corresponding to a region within amino acids 260 and 582 of HPA2 (Uniprot ID#Q8WWQ2).
Specifications
| HPA2 | |
| Polyclonal | |
| Unconjugated | |
| Q8WWQ2, B2RY83 | |
| HPSE2 | |
| Recombinant fragment corresponding to a region within amino acids 260 and 582 of HPA2 (Uniprot ID#Q8WWQ2) | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1mg/mL | |
| HPSE2 | |
| HPA2, HPR2, UFS, heparanase 3, heparanase-2, heparanase-like protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 545291, 60495 | |
| Store at -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction