Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hydroxyprostaglandin dehydrogenase 15-(NAD), Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89005564 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-005-564 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-005-564 Supplier Abnova Corporation Supplier No. H00003248B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human HPGD protein.

This gene encodes a member of the short-chain nonmetalloenzyme alcohol dehydrogenase protein family. The encoded enzyme is responsible for the metabolism of prostaglandins, which function in a variety of physiologic and cellular processes such as inflammation. Mutations in this gene result in primary autosomal recessive hypertrophic osteoarthropathy and cranioosteoarthropathy. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: MHVNGKVALVTGAAQGIGRAFAEALLLKGAKVALVDWNLEAGVQCKAALDEQFEPQKTLFIQCDVADQQQLRDTFRKVVDHFGRLDILVNNAGVNNEKNWEKTLQINLVSVISGTYLGLDYMSKQNGGEGGIIINMSSLAGLMPVAQQPVYCASKHGIVGFTRSAALAANLMNSGVRLNAICPGFVNTAILESIEKEENMGQYIEYKDHIKDMIKYYGILDPPLIANGLITLIEDDALNGAIMKITTSKGIHFQDYDTTPFQAKTQ

Specifications

Antigen hydroxyprostaglandin dehydrogenase 15-(NAD)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human HPGD protein.
Formulation No additive
Gene HPGD
Gene Accession No. BC018986
Gene Alias 15-PGDH/PGDH/PGDH1/SDR36C1
Gene Symbols HPGD
Host Species Mouse
Immunogen HPGD (AAH18986.1, 1 a.a. ∼ 266 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 3248
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.