Learn More
hydroxyprostaglandin dehydrogenase 15-(NAD), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human HPGD protein.
Supplier: Abnova Corporation H00003248D01P
Description
This gene encodes a member of the short-chain nonmetalloenzyme alcohol dehydrogenase protein family. The encoded enzyme is responsible for the metabolism of prostaglandins, which function in a variety of physiologic and cellular processes such as inflammation. Mutations in this gene result in primary autosomal recessive hypertrophic osteoarthropathy and cranioosteoarthropathy. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: MHVNGKVALVTGAAQGIGRAFAEALLLKGAKVALVDWNLEAGVQCKAALDEQFEPQKTLFIQCDVADQQQLRDTFRKVVDHFGRLDILVNNAGVNNEKNWEKTLQINLVSVISGTYLGLDYMSKQNGGEGGIIINMSSLAGLMPVAQQPVYCASKHGIVGFTRSAALAANLMNSGVRLNAICPGFVNTAILESIEKEENMGQYIEYKDHIKDMIKYYGILDPPLIANGLITLIEDDALNGAIMKITTSKGIHFQDYDTTPFQAKTQSpecifications
hydroxyprostaglandin dehydrogenase 15-(NAD) | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human HPGD protein. | |
HPGD | |
15-PGDH/PGDH/PGDH1/SDR36C1 | |
Rabbit | |
Affinity Purified | |
RUO | |
3248 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC018986 | |
HPGD | |
HPGD (AAH18986.1, 1 a.a. ∼ 266 a.a) full-length human protein. | |
100 μg | |
Primary | |
Human | |
Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.